Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610513 MIM: 615776 MIM: 156790 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP13A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP13A2 - ATPase 13A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CROCC - ciliary rootlet coiled-coil, rootletin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFAP2 - microfibrillar associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135247.1 | Intron | NP_001128719.1 | ||||
NM_001135248.1 | Intron | NP_001128720.1 | ||||
NM_002403.3 | Intron | NP_002394.1 | ||||
NM_017459.2 | Intron | NP_059453.1 | ||||
XM_011541476.2 | Intron | XP_011539778.1 |