Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCAGGTAAGAAATGTTAGTAAAT[A/T]GGCATCTAGTGGGGAGAGGGGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 145505 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACSM3 - acyl-CoA synthetase medium-chain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005622.3 | Intron | NP_005613.2 | ||||
NM_202000.2 | Intron | NP_973729.1 | ||||
XM_011545911.1 | Intron | XP_011544213.1 | ||||
XM_017023523.1 | Intron | XP_016879012.1 | ||||
XM_017023524.1 | Intron | XP_016879013.1 |
ERI2 - ERI1 exoribonuclease family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142725.1 | Intron | NP_001136197.1 | ||||
NM_080663.2 | Intron | NP_542394.2 |
Set Membership: |
HapMap |