Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604318 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2IRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GTF2IRD1 - GTF2I repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199207.1 | Intron | NP_001186136.1 | ||||
NM_005685.3 | Intron | NP_005676.3 | ||||
NM_016328.2 | Intron | NP_057412.1 | ||||
XM_006716182.3 | Intron | XP_006716245.1 | ||||
XM_006716183.3 | Intron | XP_006716246.1 | ||||
XM_011516713.1 | Intron | XP_011515015.1 | ||||
XM_017012802.1 | Intron | XP_016868291.1 | ||||
XM_017012803.1 | Intron | XP_016868292.1 | ||||
XM_017012804.1 | Intron | XP_016868293.1 | ||||
XM_017012805.1 | Intron | XP_016868294.1 |
Set Membership: |
HapMap |