Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGTGGGGTGGAAGACATCTCAT[C/T]CTTCAAAAGTCTGGTGAGGCCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612693 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
URM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
URM1 - ubiquitin related modifier 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135947.2 | Intron | NP_001129419.1 | ||||
NM_001265582.1 | Intron | NP_001252511.1 | ||||
NM_030914.3 | Intron | NP_112176.1 |