Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607576 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CECR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CECR2 - CECR2, histone acetyl-lysine reader | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290046.1 | Intron | NP_001276975.1 | ||||
NM_001290047.1 | Intron | NP_001276976.1 | ||||
XM_006724079.3 | Intron | XP_006724142.1 | ||||
XM_011546128.2 | Intron | XP_011544430.1 | ||||
XM_011546129.2 | Intron | XP_011544431.1 | ||||
XM_011546132.2 | Intron | XP_011544434.1 | ||||
XM_017028785.1 | Intron | XP_016884274.1 |
Set Membership: |
HapMap |