Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCTGAAACTTTAGAACTGCACC[A/G]ATGTCTAAGACTTTCATCCTCCTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604297 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAXBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PAXBP1 - PAX3 and PAX7 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAXBP1-AS1 - PAXBP1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNJ1 - synaptojanin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |