Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146770 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRICH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRICH1 - aspartate rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016449.3 | 1592 | Intron | NP_057533.2 | |||
XM_011530206.1 | 1592 | UTR 3 | XP_011528508.1 | |||
XM_011530207.2 | 1592 | Intron | XP_011528509.1 | |||
XM_011530208.2 | 1592 | Intron | XP_011528510.1 | |||
XM_011530209.2 | 1592 | Intron | XP_011528511.1 | |||
XM_011530210.2 | 1592 | Intron | XP_011528512.1 | |||
XM_017028813.1 | 1592 | Intron | XP_016884302.1 |
IGLL1 - immunoglobulin lambda like polypeptide 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020070.3 | 1592 | Intron | NP_064455.1 | |||
NM_152855.2 | 1592 | Intron | NP_690594.1 | |||
XM_011530169.2 | 1592 | Intron | XP_011528471.1 |