Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604050 MIM: 602142 | ||||||||||||||||||||
Literature Links: |
DLEC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DLEC1 - deleted in lung and esophageal cancer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321153.1 | 217 | Missense Mutation | CTC,TTC | L,F 66 | NP_001308082.1 | |
NM_007335.3 | 217 | Missense Mutation | CTC,TTC | L,F 66 | NP_031361.2 | |
NM_007337.3 | 217 | Missense Mutation | CTC,TTC | L,F 66 | NP_031363.2 |
PLCD1 - phospholipase C delta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |