Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGGATGCTTCAGAGCACATGGA[C/T]GATACCCAGGTCAGCACGGTGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609228 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102723521 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC102723521 - uncharacterized LOC102723521 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT3 - nudix hydrolase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006703.3 | 2024 | UTR 3 | NP_006694.1 |
RPS10-NUDT3 - RPS10-NUDT3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202470.2 | 2024 | UTR 3 | NP_001189399.1 |
Set Membership: |
HapMap Validated |