Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604646 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP50 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUP50 - nucleoporin 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007172.3 | Intron | NP_009103.2 | ||||
NM_153645.2 | Intron | NP_705931.1 | ||||
XM_005261312.1 | Intron | XP_005261369.1 | ||||
XM_005261314.1 | Intron | XP_005261371.1 | ||||
XM_006724103.1 | Intron | XP_006724166.1 | ||||
XM_006724104.1 | Intron | XP_006724167.1 | ||||
XM_011529833.1 | Intron | XP_011528135.1 | ||||
XM_017028537.1 | Intron | XP_016884026.1 | ||||
XM_017028538.1 | Intron | XP_016884027.1 |
NUP50-AS1 - NUP50 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |