Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAATTTACTTGGATAACATCAT[C/T]GGGTCATTGAAACTTTGTGTGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609405 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP8 - Rho GTPase activating protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017526.1 | Intron | NP_001017526.1 | ||||
NM_001198726.1 | Intron | NP_001185655.1 | ||||
NM_181335.2 | Intron | NP_851852.2 |
PRR5-ARHGAP8 - PRR5-ARHGAP8 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181334.5 | Intron | NP_851851.3 |
Set Membership: |
HapMap |