Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604518 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
GRAP2 - GRB2-related adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291824.1 | Intron | NP_001278753.1 | ||||
NM_001291825.1 | Intron | NP_001278754.1 | ||||
NM_001291826.1 | Intron | NP_001278755.1 | ||||
NM_001291828.1 | Intron | NP_001278757.1 | ||||
NM_004810.3 | Intron | NP_004801.1 | ||||
XM_006724376.3 | Intron | XP_006724439.1 |