Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180980 MIM: 614720 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AMD1 - adenosylmethionine decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287214.1 | Intron | NP_001274143.1 | ||||
NM_001287215.1 | Intron | NP_001274144.1 | ||||
NM_001287216.1 | Intron | NP_001274145.1 | ||||
NM_001634.5 | Intron | NP_001625.2 |
CDK19 - cyclin dependent kinase 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300960.1 | Intron | NP_001287889.1 | ||||
NM_001300963.1 | Intron | NP_001287892.1 | ||||
NM_001300964.1 | Intron | NP_001287893.1 | ||||
NM_015076.4 | Intron | NP_055891.1 | ||||
XM_005266871.3 | Intron | XP_005266928.1 | ||||
XM_011535630.2 | Intron | XP_011533932.1 | ||||
XM_011535631.2 | Intron | XP_011533933.1 | ||||
XM_011535632.1 | Intron | XP_011533934.1 | ||||
XM_017010587.1 | Intron | XP_016866076.1 | ||||
XM_017010588.1 | Intron | XP_016866077.1 |
Set Membership: |
Validated |