Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609898 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf31 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C22orf31 - chromosome 22 open reading frame 31 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KREMEN1 - kringle containing transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039570.2 | Intron | NP_001034659.2 | ||||
NM_032045.4 | Intron | NP_114434.3 | ||||
XM_011530429.2 | Intron | XP_011528731.1 | ||||
XM_011530430.2 | Intron | XP_011528732.1 | ||||
XM_011530431.2 | Intron | XP_011528733.1 | ||||
XM_011530432.2 | Intron | XP_011528734.1 | ||||
XM_017028989.1 | Intron | XP_016884478.1 |
Set Membership: |
HapMap |