Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
66 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604743 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDAH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DDAH1 - dimethylarginine dimethylaminohydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134445.1 | 2403 | UTR 3 | NP_001127917.1 | |||
NM_012137.3 | 2403 | UTR 3 | NP_036269.1 | |||
XM_005270707.2 | 2403 | UTR 3 | XP_005270764.1 | |||
XM_005270710.2 | 2403 | UTR 3 | XP_005270767.1 | |||
XM_011541158.1 | 2403 | UTR 3 | XP_011539460.1 | |||
XM_017000889.1 | 2403 | UTR 3 | XP_016856378.1 | |||
XM_017000890.1 | 2403 | UTR 3 | XP_016856379.1 |
Set Membership: |
HapMap JSNP |