Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611317 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIK3R5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PIK3R5 - phosphoinositide-3-kinase regulatory subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142633.2 | Intron | NP_001136105.1 | ||||
NM_001251851.1 | Intron | NP_001238780.1 | ||||
NM_001251852.1 | Intron | NP_001238781.1 | ||||
NM_001251853.1 | Intron | NP_001238782.1 | ||||
NM_001251855.1 | Intron | NP_001238784.1 | ||||
NM_014308.3 | Intron | NP_055123.2 | ||||
XM_005256580.3 | Intron | XP_005256637.1 |
Set Membership: |
HapMap |