Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600428 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VAV2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
VAV2 - vav guanine nucleotide exchange factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134398.1 | Intron | NP_001127870.1 | ||||
NM_003371.3 | Intron | NP_003362.2 | ||||
XM_005272213.1 | Intron | XP_005272270.1 | ||||
XM_017015108.1 | Intron | XP_016870597.1 | ||||
XM_017015109.1 | Intron | XP_016870598.1 | ||||
XM_017015110.1 | Intron | XP_016870599.1 | ||||
XM_017015111.1 | Intron | XP_016870600.1 | ||||
XM_017015112.1 | Intron | XP_016870601.1 | ||||
XM_017015113.1 | Intron | XP_016870602.1 |