Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116790 MIM: 606448 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COMT - catechol-O-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000754.3 | Intron | NP_000745.1 | ||||
NM_001135161.1 | Intron | NP_001128633.1 | ||||
NM_001135162.1 | Intron | NP_001128634.1 | ||||
NM_007310.2 | Intron | NP_009294.1 | ||||
XM_011529886.1 | Intron | XP_011528188.1 | ||||
XM_017028594.1 | Intron | XP_016884083.1 | ||||
XM_017028595.1 | Intron | XP_016884084.1 |
TXNRD2 - thioredoxin reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282512.1 | Intron | NP_001269441.1 | ||||
NM_006440.4 | Intron | NP_006431.2 |
Set Membership: |
HapMap |