Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCACCAGCTTGGTGCCGCCCT[A/G]CTCCAGGTGCACCGTGGCCGAGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600725 | ||||||||||||||||||||
Literature Links: |
SHH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHH - sonic hedgehog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000193.3 | 3574 | Nonsense Mutation | CAG,TAG | Q,* 209 | NP_000184.1 | |
NM_001310462.1 | 3574 | Intron | NP_001297391.1 | |||
XM_011516479.2 | 3574 | Nonsense Mutation | CAG,TAG | Q,* 122 | XP_011514781.1 | |
XM_011516480.2 | 3574 | Nonsense Mutation | CAG,TAG | Q,* 122 | XP_011514782.1 |