Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300103 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHROOM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SHROOM2 - shroom family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320663.1 | Intron | NP_001307592.1 | ||||
NM_001320664.1 | Intron | NP_001307593.1 | ||||
NM_001649.3 | Intron | NP_001640.1 | ||||
XM_005274500.4 | Intron | XP_005274557.2 | ||||
XM_011545506.1 | Intron | XP_011543808.1 | ||||
XM_017029492.1 | Intron | XP_016884981.1 | ||||
XM_017029493.1 | Intron | XP_016884982.1 | ||||
XM_017029494.1 | Intron | XP_016884983.1 |
Set Membership: |
HapMap Validated |