Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613138 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TSPAN12 - tetraspanin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012338.3 | 646 | Silent Mutation | CCG,CCT | P,P 255 | NP_036470.1 | |
XM_005250239.2 | 646 | Silent Mutation | CCG,CCT | P,P 255 | XP_005250296.1 | |
XM_011515993.1 | 646 | Silent Mutation | CCG,CCT | P,P 255 | XP_011514295.1 | |
XM_011515994.1 | 646 | Silent Mutation | CCG,CCT | P,P 255 | XP_011514296.1 | |
XM_017011912.1 | 646 | Silent Mutation | CCG,CCT | P,P 255 | XP_016867401.1 | |
XM_017011913.1 | 646 | Silent Mutation | CCG,CCT | P,P 230 | XP_016867402.1 | |
XM_017011914.1 | 646 | Silent Mutation | CCG,CCT | P,P 128 | XP_016867403.1 |