Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614730 MIM: 608292 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM214B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM214B - family with sequence similarity 214 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGO - phosphatidylinositol glycan anchor biosynthesis class O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STOML2 - stomatin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287031.1 | Intron | NP_001273960.1 | ||||
NM_001287032.1 | Intron | NP_001273961.1 | ||||
NM_001287033.1 | Intron | NP_001273962.1 | ||||
NM_013442.2 | Intron | NP_038470.1 |
Set Membership: |
HapMap |