Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147890 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MX2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MX2 - MX dynamin like GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002463.1 | 88 | Intron | NP_002454.1 | |||
XM_005260983.4 | 88 | UTR 5 | XP_005261040.1 | |||
XM_005260984.1 | 88 | Intron | XP_005261041.1 | |||
XM_011529571.1 | 88 | Intron | XP_011527873.1 | |||
XM_011529572.1 | 88 | Intron | XP_011527874.1 | |||
XM_011529573.1 | 88 | Intron | XP_011527875.1 | |||
XM_011529574.2 | 88 | Intron | XP_011527876.1 |
Set Membership: |
HapMap Validated |