Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGTCTGTAGTTTTCACTTCCTC[A/C]CCTTCAAGAGGAAGATGATGCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605568 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927550 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101927550 - uncharacterized LOC101927550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMURF1 - SMAD specific E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199847.1 | 4266 | UTR 3 | NP_001186776.1 | |||
NM_020429.2 | 4266 | UTR 3 | NP_065162.1 | |||
NM_181349.2 | 4266 | UTR 3 | NP_851994.1 | |||
XM_017012457.1 | 4266 | UTR 3 | XP_016867946.1 |
Set Membership: |
HapMap |