Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGGCAGCGCCCCACCTGCACCG[A/T]CTCGCGGTAGAACGCGGGCACCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600856 MIM: 603240 | ||||||||||||||||||||
Literature Links: |
CDKN1C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDKN1C - cyclin dependent kinase inhibitor 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000076.2 | 418 | Missense Mutation | ACG,TCG | T,S 94 | NP_000067.1 | |
NM_001122630.1 | 418 | Missense Mutation | ACG,TCG | T,S 83 | NP_001116102.1 | |
NM_001122631.1 | 418 | Missense Mutation | ACG,TCG | T,S 83 | NP_001116103.1 | |
XM_005252732.4 | 418 | Missense Mutation | ACG,TCG | T,S 83 | XP_005252789.1 | |
XM_017017088.1 | 418 | Missense Mutation | ACG,TCG | T,S 94 | XP_016872577.1 |
SLC22A18AS - solute carrier family 22 member 18 antisense | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |