Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614133 | ||||||||||||||||||||
Literature Links: |
HEPACAM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEPACAM2 - HEPACAM family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039372.2 | Intron | NP_001034461.1 | ||||
NM_001288804.1 | Intron | NP_001275733.1 | ||||
NM_001288810.1 | Intron | NP_001275739.1 | ||||
NM_198151.2 | Intron | NP_937794.1 | ||||
XM_011516000.2 | Intron | XP_011514302.1 | ||||
XM_011516001.1 | Intron | XP_011514303.1 |