Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601273 MIM: 190315 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLTCL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLTCL1 - clathrin heavy chain like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001835.3 | 3350 | UTR 3 | NP_001826.3 | |||
NM_007098.3 | 3350 | UTR 3 | NP_009029.3 | |||
XM_011530401.2 | 3350 | UTR 3 | XP_011528703.1 | |||
XM_017028953.1 | 3350 | UTR 3 | XP_016884442.1 | |||
XM_017028954.1 | 3350 | UTR 3 | XP_016884443.1 | |||
XM_017028955.1 | 3350 | UTR 3 | XP_016884444.1 | |||
XM_017028956.1 | 3350 | UTR 3 | XP_016884445.1 | |||
XM_017028957.1 | 3350 | Intron | XP_016884446.1 |
LINC01311 - long intergenic non-protein coding RNA 1311 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A1 - solute carrier family 25 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |