Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 158343 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ABCC1 - ATP binding cassette subfamily C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004996.3 | Intron | NP_004987.2 | ||||
XM_011522497.1 | Intron | XP_011520799.1 | ||||
XM_011522498.2 | Intron | XP_011520800.1 | ||||
XM_017023237.1 | Intron | XP_016878726.1 | ||||
XM_017023238.1 | Intron | XP_016878727.1 | ||||
XM_017023239.1 | Intron | XP_016878728.1 | ||||
XM_017023240.1 | Intron | XP_016878729.1 | ||||
XM_017023241.1 | Intron | XP_016878730.1 | ||||
XM_017023242.1 | Intron | XP_016878731.1 | ||||
XM_017023243.1 | Intron | XP_016878732.1 |
Set Membership: |
HapMap |