Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602857 MIM: 609780 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHN2 - chimerin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039936.2 | Intron | NP_001035025.1 | ||||
NM_001293069.1 | Intron | NP_001279998.1 | ||||
NM_001293070.1 | Intron | NP_001279999.1 | ||||
NM_001293071.1 | Intron | NP_001280000.1 | ||||
NM_001293072.1 | Intron | NP_001280001.1 | ||||
NM_001293073.1 | Intron | NP_001280002.1 | ||||
NM_001293075.1 | Intron | NP_001280004.1 | ||||
NM_001293076.1 | Intron | NP_001280005.1 | ||||
NM_001293077.1 | Intron | NP_001280006.1 | ||||
NM_001293078.1 | Intron | NP_001280007.1 | ||||
NM_001293079.1 | Intron | NP_001280008.1 | ||||
NM_001293080.1 | Intron | NP_001280009.1 | ||||
NM_001293081.1 | Intron | NP_001280010.1 | ||||
NM_004067.3 | Intron | NP_004058.1 | ||||
XM_011515105.2 | Intron | XP_011513407.1 | ||||
XM_011515106.2 | Intron | XP_011513408.1 | ||||
XM_011515107.2 | Intron | XP_011513409.1 | ||||
XM_017011721.1 | Intron | XP_016867210.1 | ||||
XM_017011722.1 | Intron | XP_016867211.1 |
CPVL - carboxypeptidase, vitellogenic like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |