Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGGGAGGCAACTGGACCCTAG[G/T]GTCTGGACAGGCCGTGGCAGGTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104620 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD14A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ABHD14A - abhydrolase domain containing 14A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015407.4 | Intron | NP_056222.2 |
ABHD14A-ACY1 - ABHD14A-ACY1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316331.1 | Intron | NP_001303260.1 |
ABHD14B - abhydrolase domain containing 14B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ACY1 - aminoacylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |