Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603049 | ||||||||||||||||||||
Literature Links: |
TXNL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TXNL1 - thioredoxin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004786.2 | Intron | NP_004777.1 | ||||
XM_006722580.3 | Intron | XP_006722643.1 | ||||
XM_011526263.2 | Intron | XP_011524565.1 | ||||
XM_011526264.2 | Intron | XP_011524566.1 | ||||
XM_017026093.1 | Intron | XP_016881582.1 | ||||
XM_017026094.1 | Intron | XP_016881583.1 |