Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603049 MIM: 613473 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TXNL1 - thioredoxin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004786.2 | Intron | NP_004777.1 | ||||
XM_006722580.3 | Intron | XP_006722643.1 | ||||
XM_011526263.2 | Intron | XP_011524565.1 | ||||
XM_011526264.2 | Intron | XP_011524566.1 | ||||
XM_017026093.1 | Intron | XP_016881582.1 | ||||
XM_017026094.1 | Intron | XP_016881583.1 |
WDR7 - WD repeat domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015285.2 | Intron | NP_056100.2 | ||||
NM_052834.2 | Intron | NP_443066.2 | ||||
XM_006722431.2 | Intron | XP_006722494.1 | ||||
XM_011525888.1 | Intron | XP_011524190.1 | ||||
XM_017025682.1 | Intron | XP_016881171.1 | ||||
XM_017025683.1 | Intron | XP_016881172.1 |
Set Membership: |
HapMap |