Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616372 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCAF4 - DDB1 and CUL4 associated factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163508.1 | 102 | Intron | NP_001156980.1 | |||
NM_001163509.1 | 102 | Intron | NP_001156981.1 | |||
NM_015604.3 | 102 | Intron | NP_056419.2 | |||
NM_181340.2 | 102 | Intron | NP_851937.1 | |||
NM_181341.2 | 102 | Intron | NP_851938.2 | |||
XM_017021201.1 | 102 | Intron | XP_016876690.1 | |||
XM_017021202.1 | 102 | Intron | XP_016876691.1 | |||
XM_017021203.1 | 102 | Intron | XP_016876692.1 | |||
XM_017021204.1 | 102 | Intron | XP_016876693.1 | |||
XM_017021205.1 | 102 | UTR 5 | XP_016876694.1 | |||
XM_017021206.1 | 102 | UTR 5 | XP_016876695.1 | |||
XM_017021207.1 | 102 | Intron | XP_016876696.1 | |||
XM_017021208.1 | 102 | Intron | XP_016876697.1 | |||
XM_017021209.1 | 102 | Intron | XP_016876698.1 | |||
XM_017021210.1 | 102 | Intron | XP_016876699.1 | |||
XM_017021211.1 | 102 | Intron | XP_016876700.1 | |||
XM_017021212.1 | 102 | UTR 5 | XP_016876701.1 | |||
XM_017021213.1 | 102 | UTR 5 | XP_016876702.1 | |||
XM_017021214.1 | 102 | Intron | XP_016876703.1 | |||
XM_017021215.1 | 102 | Intron | XP_016876704.1 |