Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35F4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC35F4 - solute carrier family 35 member F4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206920.1 | Intron | NP_001193849.1 | ||||
NM_001306087.1 | Intron | NP_001293016.1 | ||||
XM_011536720.2 | Intron | XP_011535022.1 | ||||
XM_011536721.2 | Intron | XP_011535023.1 | ||||
XM_011536723.2 | Intron | XP_011535025.1 | ||||
XM_011536724.2 | Intron | XP_011535026.1 | ||||
XM_011536725.1 | Intron | XP_011535027.1 | ||||
XM_017021258.1 | Intron | XP_016876747.1 | ||||
XM_017021259.1 | Intron | XP_016876748.1 | ||||
XM_017021260.1 | Intron | XP_016876749.1 | ||||
XM_017021261.1 | Intron | XP_016876750.1 |