Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605211 MIM: 616533 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BARHL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BARHL1 - BarH like homeobox 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DDX31 - DEAD-box helicase 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322340.1 | 2666 | UTR 3 | NP_001309269.1 | |||
NM_001322341.1 | 2666 | UTR 3 | NP_001309270.1 | |||
NM_001322342.1 | 2666 | UTR 3 | NP_001309271.1 | |||
NM_001322343.1 | 2666 | UTR 3 | NP_001309272.1 | |||
NM_001322344.1 | 2666 | Intron | NP_001309273.1 | |||
NM_022779.8 | 2666 | UTR 3 | NP_073616.6 | |||
NM_138620.1 | 2666 | Intron | NP_619526.1 | |||
XM_005272207.4 | 2666 | UTR 3 | XP_005272264.1 | |||
XM_006717236.3 | 2666 | UTR 3 | XP_006717299.1 | |||
XM_011518921.2 | 2666 | Intron | XP_011517223.1 | |||
XM_011518922.2 | 2666 | Intron | XP_011517224.1 |