Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118946 | ||||||||||||||||||||
Literature Links: |
CNTFR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNTFR - ciliary neurotrophic factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207011.1 | Intron | NP_001193940.1 | ||||
NM_001842.4 | Intron | NP_001833.1 | ||||
NM_147164.2 | Intron | NP_671693.1 | ||||
XM_017014260.1 | Intron | XP_016869749.1 | ||||
XM_017014261.1 | Intron | XP_016869750.1 | ||||
XM_017014262.1 | Intron | XP_016869751.1 | ||||
XM_017014263.1 | Intron | XP_016869752.1 | ||||
XM_017014264.1 | Intron | XP_016869753.1 | ||||
XM_017014265.1 | Intron | XP_016869754.1 |