Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607451 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMEB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GMEB2 - glucocorticoid modulatory element binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012384.4 | 1280 | Silent Mutation | CTA,CTG | L,L 475 | NP_036516.1 | |
XM_005260202.4 | 1280 | Silent Mutation | CTA,CTG | L,L 475 | XP_005260259.1 | |
XM_006723776.2 | 1280 | Silent Mutation | CTA,CTG | L,L 397 | XP_006723839.1 | |
XM_011528779.2 | 1280 | Silent Mutation | CTA,CTG | L,L 397 | XP_011527081.1 |