Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM217 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM217 - transmembrane protein 217 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162900.1 | 867 | Intron | NP_001156372.1 | |||
NM_001286401.1 | 867 | Intron | NP_001273330.1 | |||
NM_145316.3 | 867 | Intron | NP_660359.2 | |||
XM_011514362.1 | 867 | Intron | XP_011512664.1 | |||
XM_011514363.2 | 867 | UTR 3 | XP_011512665.1 | |||
XM_011514364.2 | 867 | Intron | XP_011512666.1 | |||
XM_011514365.2 | 867 | Intron | XP_011512667.1 | |||
XM_011514366.2 | 867 | UTR 3 | XP_011512668.1 | |||
XM_011514367.2 | 867 | Intron | XP_011512669.1 | |||
XM_011514369.2 | 867 | Intron | XP_011512671.1 | |||
XM_011514370.2 | 867 | Intron | XP_011512672.1 |