Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609205 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAB2IP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DAB2IP - DAB2 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032552.3 | Intron | NP_115941.2 | ||||
NM_138709.2 | Intron | NP_619723.1 | ||||
XM_005251719.4 | Intron | XP_005251776.1 | ||||
XM_005251721.1 | Intron | XP_005251778.1 | ||||
XM_005251724.4 | Intron | XP_005251781.1 | ||||
XM_011518264.2 | Intron | XP_011516566.1 | ||||
XM_011518265.2 | Intron | XP_011516567.1 | ||||
XM_011518266.2 | Intron | XP_011516568.1 | ||||
XM_011518267.2 | Intron | XP_011516569.1 | ||||
XM_011518270.2 | Intron | XP_011516572.1 | ||||
XM_011518271.2 | Intron | XP_011516573.1 | ||||
XM_017014298.1 | Intron | XP_016869787.1 | ||||
XM_017014299.1 | Intron | XP_016869788.1 | ||||
XM_017014300.1 | Intron | XP_016869789.1 |
Set Membership: |
HapMap |