Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
51 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615125 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD13C - ankyrin repeat domain 13C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030816.4 | Intron | NP_110443.3 | ||||
XM_005271234.3 | Intron | XP_005271291.1 | ||||
XM_005271235.3 | Intron | XP_005271292.1 | ||||
XM_006710929.3 | Intron | XP_006710992.1 | ||||
XM_017002413.1 | Intron | XP_016857902.1 | ||||
XM_017002414.1 | Intron | XP_016857903.1 | ||||
XM_017002415.1 | Intron | XP_016857904.1 | ||||
XM_017002416.1 | Intron | XP_016857905.1 |
Set Membership: |
HapMap |