Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602927 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR19 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
GPR19 - G protein-coupled receptor 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006143.2 | 5200 | Intron | NP_006134.1 | |||
XM_011520623.2 | 5200 | UTR 3 | XP_011518925.1 | |||
XM_011520624.2 | 5200 | UTR 3 | XP_011518926.1 | |||
XM_011520625.2 | 5200 | UTR 3 | XP_011518927.1 | |||
XM_017019212.1 | 5200 | UTR 3 | XP_016874701.1 | |||
XM_017019213.1 | 5200 | UTR 3 | XP_016874702.1 | |||
XM_017019214.1 | 5200 | UTR 3 | XP_016874703.1 | |||
XM_017019215.1 | 5200 | UTR 3 | XP_016874704.1 | |||
XM_017019216.1 | 5200 | UTR 3 | XP_016874705.1 |
Set Membership: |
HapMap |