Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600241 MIM: 605875 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GPR3 - G protein-coupled receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005281.3 | 764 | Missense Mutation | CAC,CGC | H,R 222 | NP_005272.1 |
WASF2 - WAS protein family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |