Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607981 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUB1 - negative regulator of ubiquitin like proteins 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243351.1 | Intron | NP_001230280.1 | ||||
NM_016118.4 | Intron | NP_057202.3 | ||||
XM_005250011.2 | Intron | XP_005250068.1 | ||||
XM_017012304.1 | Intron | XP_016867793.1 | ||||
XM_017012305.1 | Intron | XP_016867794.1 | ||||
XM_017012306.1 | Intron | XP_016867795.1 | ||||
XM_017012307.1 | Intron | XP_016867796.1 | ||||
XM_017012308.1 | Intron | XP_016867797.1 | ||||
XM_017012309.1 | Intron | XP_016867798.1 | ||||
XM_017012310.1 | Intron | XP_016867799.1 | ||||
XM_017012311.1 | Intron | XP_016867800.1 | ||||
XM_017012312.1 | Intron | XP_016867801.1 | ||||
XM_017012313.1 | Intron | XP_016867802.1 | ||||
XM_017012314.1 | Intron | XP_016867803.1 |