Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616303 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR91 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WDR91 - WD repeat domain 91 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014149.3 | Intron | NP_054868.3 | ||||
XM_011516083.2 | Intron | XP_011514385.1 | ||||
XM_011516086.2 | Intron | XP_011514388.1 | ||||
XM_017012070.1 | Intron | XP_016867559.1 | ||||
XM_017012071.1 | Intron | XP_016867560.1 | ||||
XM_017012072.1 | Intron | XP_016867561.1 |