Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609987 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STRA8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
STRA8 - stimulated by retinoic acid 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182489.1 | 218 | Intron | NP_872295.1 | |||
XM_011516137.2 | 218 | UTR 5 | XP_011514439.1 | |||
XM_011516138.2 | 218 | Intron | XP_011514440.1 | |||
XM_011516139.2 | 218 | UTR 5 | XP_011514441.1 | |||
XM_017012150.1 | 218 | UTR 5 | XP_016867639.1 |