Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
62 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC5A9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SLC5A9 - solute carrier family 5 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011547.2 | Intron | NP_001011547.2 | ||||
NM_001135181.1 | Intron | NP_001128653.1 | ||||
XM_011540924.2 | Intron | XP_011539226.1 | ||||
XM_011540925.2 | Intron | XP_011539227.1 | ||||
XM_011540926.2 | Intron | XP_011539228.1 | ||||
XM_011540927.1 | Intron | XP_011539229.1 | ||||
XM_011540928.1 | Intron | XP_011539230.1 | ||||
XM_011540929.2 | Intron | XP_011539231.1 | ||||
XM_017000558.1 | Intron | XP_016856047.1 |
Set Membership: |
HapMap |