Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602851 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADGRV1 - adhesion G protein-coupled receptor V1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032119.3 | Intron | NP_115495.3 | ||||
XM_017009963.1 | Intron | XP_016865452.1 | ||||
XM_017009964.1 | Intron | XP_016865453.1 | ||||
XM_017009965.1 | Intron | XP_016865454.1 | ||||
XM_017009966.1 | Intron | XP_016865455.1 | ||||
XM_017009967.1 | Intron | XP_016865456.1 | ||||
XM_017009968.1 | Intron | XP_016865457.1 | ||||
XM_017009969.1 | Intron | XP_016865458.1 | ||||
XM_017009970.1 | Intron | XP_016865459.1 | ||||
XM_017009971.1 | Intron | XP_016865460.1 | ||||
XM_017009972.1 | Intron | XP_016865461.1 | ||||
XM_017009973.1 | Intron | XP_016865462.1 | ||||
XM_017009974.1 | Intron | XP_016865463.1 |
Set Membership: |
HapMap Validated |