Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142810 MIM: 600783 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HARS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HARS - histidyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258040.2 | Intron | NP_001244969.1 | ||||
NM_001258041.2 | Intron | NP_001244970.1 | ||||
NM_001258042.2 | Intron | NP_001244971.1 | ||||
NM_001289092.1 | Intron | NP_001276021.1 | ||||
NM_001289093.1 | Intron | NP_001276022.1 | ||||
NM_001289094.1 | Intron | NP_001276023.1 | ||||
NM_002109.5 | Intron | NP_002100.2 |
HARS2 - histidyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278731.1 | Intron | NP_001265660.1 | ||||
NM_001278732.1 | Intron | NP_001265661.1 | ||||
NM_012208.3 | Intron | NP_036340.1 | ||||
XM_011537619.1 | Intron | XP_011535921.1 | ||||
XM_011537620.1 | Intron | XP_011535922.1 | ||||
XM_017009287.1 | Intron | XP_016864776.1 | ||||
XM_017009288.1 | Intron | XP_016864777.1 | ||||
XM_017009289.1 | Intron | XP_016864778.1 | ||||
XM_017009290.1 | Intron | XP_016864779.1 | ||||
XM_017009291.1 | Intron | XP_016864780.1 | ||||
XM_017009292.1 | Intron | XP_016864781.1 |
ZMAT2 - zinc finger matrin-type 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |