Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTTCCCCGAGGAGCCGCTGCCCC[C/G]GGGACCCCCTGGCACTGTGCGCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604056 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HS3ST2 - heparan sulfate-glucosamine 3-sulfotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006043.1 | 267 | Intron | NP_006034.1 | |||
XM_011546001.2 | 267 | UTR 5 | XP_011544303.1 | |||
XM_011546002.2 | 267 | UTR 5 | XP_011544304.1 |
LOC107984851 - collagen alpha-1(XVI) chain-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017023983.1 | 267 | Missense Mutation | CGG,GGG | R,G 53 | XP_016879472.1 |