Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604557 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF423 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF423 - zinc finger protein 423 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271620.2 | 6081 | UTR 3 | NP_001258549.1 | |||
NM_015069.4 | 6081 | UTR 3 | NP_055884.2 | |||
XM_005255856.4 | 6081 | Intron | XP_005255913.1 | |||
XM_005255857.4 | 6081 | Intron | XP_005255914.1 | |||
XM_006721171.3 | 6081 | UTR 3 | XP_006721234.1 | |||
XM_017023076.1 | 6081 | UTR 3 | XP_016878565.1 | |||
XM_017023077.1 | 6081 | Intron | XP_016878566.1 | |||
XM_017023078.1 | 6081 | Intron | XP_016878567.1 |
Set Membership: |
HapMap |